missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Bovine Albumin, from New Zealand-sourced Bovine Plasma

Product Code. 17368563 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
1 kg
Unit Size:
1000g
100g
25g
500g
50g
This item is not returnable. View return policy

Product Code. 17368563

Brand: MP Biomedicals™ 0218062025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies

  • From New Zealand-sourced Bovine Plasma
  • Microbiological Grade
  • ≥97% Total Protein
  • Yellow to Brown Powder
  • Synonyms: BSA, Bovine albumin, Albumin from bovine serum
  • CAS Number: 9048-46-8
  • EC Number: 232-936-2
  • MDL Number: MFCD00130384

TRUSTED_SUSTAINABILITY

Specifications

Formulation ≥97% Total Protein
Name Bovine Albumin
Quantity 25 g
Source Bovine
Form Powder
Purity or Quality Grade Microbiological Grade
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.