missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Bovine Albumin, from New Zealand-sourced Bovine Plasma
€ 60.95 - € 2782.50
Specifications
Formulation | ≥97% Total Protein |
---|---|
Name | Bovine Albumin |
Form | Powder |
Purity or Quality Grade | Microbiological Grade |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17368563
|
MP Biomedicals™
0218062025 |
25g |
€ 108.65
25g |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17358563
|
MP Biomedicals™
0218062010 |
10g |
€ 60.95
10g |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17378563
|
MP Biomedicals™
0218062050 |
50g |
€ 204.58
50g |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17388563
|
MP Biomedicals™
0218062080 |
100g |
€ 333.90
100g |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17398563
|
MP Biomedicals™
0218062090 |
500g |
€ 1478.70
500g |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15742262
|
MP Biomedicals™
0218062091 |
1kg |
€ 2782.50
1000g |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies
- From New Zealand-sourced Bovine Plasma
- Microbiological Grade
- ≥97% Total Protein
- Yellow to Brown Powder
- Synonyms: BSA, Bovine albumin, Albumin from bovine serum
- CAS Number: 9048-46-8
- EC Number: 232-936-2
- MDL Number: MFCD00130384
Specifications
≥97% Total Protein | |
Bovine Albumin | |
Powder | |
Microbiological Grade |