missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CIRCLEGROW™ Large Capsules
Shop All MP Biomedicals Products
Click to view available options
Quantity:
454 g
1000 g
Unit Size:
1kg
454g
Description
CIRCLEGROW Medium consists of a protein and amino acid source, biological buffer, and a complex carbon source that gets metabolized at a much slower rate than glucose, thus prolonging the cell cycle and preventing cell death
during the log phase of growth. Cells grown with CIRCLEGROW Medium remain healthy for more than two days and continue to divide slowly after the log phase of the growth has been completed.
- Reduce hands-on work and preparation time
- Cultures do not need to be constantly monitored by O.D. readings due to the incremental post-log cell growth
- Obtain excellent yields of plasmid DNA from smaller culture volumes
- Confidently leave cultures overnight or over the weekend
- Capsules are the most convenient form of this unique growth media
- Pre-mixed CIRCLEGROW is also available in bulk powder, single-use pouches and sterile liquid
- Use 8 large capsules per liter;
- Contents/liter: Proprietary Formulation
Specifications
Specifications
| Certifications/Compliance | Molecular biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Dehydrated |
| Grade | Molecular biology |
| Product Type | Microbiology Media |
| Formulation | Yeast extract, Tryptone broth, Vitamins, Biological Buffers have been added for proper pH adjustment |
| For Use With (Application) | For growth of Escherichia coli, plasmid and certified for plasmid and other bacterial cultures in liquid medium |
| Quantity | 454 g |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion