Learn More
MP Biomedicals™ LymphoSep™ Lymphocyte Separation Medium
Product Code. 11454815 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 11454815
Brand: MP Biomedicals™ 091692254
LymphoSep™ Lymphocyte Separation Medium is a sterile, iso-osmotic, low viscosity solution for density centrifugation yielding a clear separation of lymphocytes from heparinized or defibrinated human blood.
  • Lymphocytes are isolated by layering diluted, heparinized blood over a solution of sodium metrizoate that contains a polysaccharide
  • Polysaccharide aggregates the erythrocytes so that they can be sedimented by low centrifugation
  • The less dense lymphocytes remain as a clearly defined band at the plasma Lymphosep interface
  • Provides:
    • In vitro isolation of lymphocytes from diluted whole blood
    • Maximum yield of mononuclear cells
    • Easy one-step centrifugation
    • >96% cell viability of lymphocytes
    • Low endotoxins
    • Autoclave sterilization
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 15° to 30°C
Product Type Lymphosep Lymphocyte Separation Medium
Sterility Sterile
pH 6 to 9
Quantity 500 mL