Learn More
MP Biomedicals™ Proteinase K
Suitable for both protein and nucleic acid isolation
Brand: MP Biomedicals™ 0219350425
73.78 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : CAS : 39450-01-6 Weight : 0.01000kg
Description
Proteinase K, [39450-01-6]
Proteolytic Activity: >30mAnson Units/mg
Unit Definition: One Anson unit is the amount of enzyme which liberates 1 μmol of Folin-positive amino acids per minute at pH 7.5 and 35°C, using hemoglobin as substrate
- From Tritirachium album
- Chromatographically purified
- Exhibits proteolytic activity on proteins, peptides, glycoproteins, amides and esters.
- Active with nitroanilides of amino acids with protected amino groups, excluding arginine.
- Useful in the isolation of DNA and RNA, in the analysis of membrane structures and protein structure.
Specifications
Protein Structure Analysis | |
Proteinase K | |
Chromatography | |
Tritirachium album Limber | |
Lyophilized |
∽30 | |
7.5 to 12 | |
25mg | |
4°C |
Safety and Handling
- Proteinase K
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.