Learn More
MP Biomedicals™ SPIN Module and Recovery Tubes
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 Pk.
60 Pk.
Unit Size:
Pack of 100
Pack of 60
Description
- Spin Module give faster and cleaner separations than traditional methods such as columns, filters, or simple centrifugation
- When used with the GENECLEAN™ or MERmaid™ protocols, they combine the speed, efficiency, and selectivity of the GLASSMILK™ binding reaction with an even more rapid execution of the separation, washing, and elution steps
Specifications
Specifications
| Content And Storage | Room Temperature |
| Sample Type | Lysate or other suspension |
| For Use With (Application) | SPIN Module is applied in molecular biology for its ability to separate particles from solutions in a few seconds. |
| Product Type | SPIN Module and Recovery Tubes |
| Quantity | 60 Pk. |
| Sample Volume | 500 to 700 μL liquid per centrifugation, Larger volumes require multiple loadings |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.