Learn More
MP Biomedicals™ TeenPrep™ Lysing Matrix Z, 15 mL Tubes
Lysing Matrix Z contains 2.0 mm diameter yttria-stabilized zirconium oxide beads and is used to grind soft animal and plant tissues and insects.
Brand:  MP Biomedicals™ 116978025
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15232860
  • € 258.64 / Pack of 25
Estimated Shipment: 31-05-2024
to see stock.
Description
Description
Lysing Matrix Z tubes can be used with FastPrep instruments or similar bead beaters. They are ideal for lysis of animal and plant tissue and whole insects. Lysing Matrix Z is an alternative to the very aggressive Lysing Matrix A for applications such as RNA and protein isolation from very tough tissue.
  • Tube size: 15 mL
  • Samples types: Animal tissue, plant tissue, tough stems, roots, seeds, whole insects
  • Matrix composition: 2 mm diameter yttria-stabilized zirconium oxide beads
Specifications
Specifications
Yttria-stabilized Zirconium Oxide
FastPrep instrument or other bead beater
Lysing Matrix Z
Animal Tissues, Plants, Insects
Nucleic Acid Isolation and Purification, Homogenization
25 x 15 mL Tubes