Learn More
MP Biomedicals™ Transport Factors
Shop All MP Biomedicals Products
Click to view available options
pH:
6.8
7.24
Unit Size:
1g
Description
Transport factors are proteins which enable small metabolites to be carried in usable form to cells. Transferrin binds iron and makes it available to cells in a recognizable form.
†Contains <1.0% iron.
Specifications
Specifications
| Content And Storage | -20°C |
| Product Type | Transferrin |
| pH | 7.24 |
| Quantity | 1 g |
| Source | Pooled Human Plasma |