missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Trypsin, 1 - 250, From Hog Pancreas

Product Code. 11453561 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
Unit Size:
100g
1kg
250g
25g
500g
This item is not returnable. View return policy

Product Code. 11453561

Brand: MP Biomedicals™ 0210313990

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

From Hog Pancreas

Trypsin, 1 - 250, [9002-07-7]

  • Parenzymol
  • Dry powder: stable
  • Protease act: min 250,000 USP units/gm

Dry Powder

TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 24000
Name Trypsin
pH Range ∽8
Quantity 500 g
Source Hog
Storage Requirements 4°C
Form Powder
compliance-icons compliance-icons
Product Identifier
  • Trypsin, 1-250
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Trypsin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.