missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPG, Mouse anti-Human, Clone: 1G6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004350-M10.100ug
This item is not returnable.
View return policy
Description
Sequence: MPARSGAQFCRRMGQKKQRPARAGQPHSSSDAAQAPAEQPHSSSDAAQAPCPRERCLGPPTTPGPYRSIYFSSPKGHLTRLGLEFFDQPASpecifications
| MPG | |
| Monoclonal | |
| Unconjugated | |
| N-methylpurine-DNA glycosylase | |
| AAG/APNG/CRA36.1/MDG/Mid1/PIG11/PIG16/anpg | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1G6 | |
| In 1x PBS, pH 7.4 | |
| BC014991 | |
| MPG | |
| MPG (AAH14991, 1 a.a. ∼ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4350 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction