missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582252
This item is not returnable.
View return policy
Description
Immunogen sequence: YFCMNISSQG DGACVLLRAL EPLEGLETMR QLRSTLRKGT ASRVLKDREL CSGPSKLCQA LAINKSFDQR DLAQDEAVWL ERGPLEPSEP AVVAAARVGV GHAGEWARKP LRFYVRGS Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 84%.
MPG functions in the hydrolysis of the deoxyribose N-glycosidic bond to excise 3-methyladenine, and 7-methylguanine from the damaged DNA polymer formed by alkylation lesions.
Specifications
| MPG | |
| Polyclonal | |
| Unconjugated | |
| MPG | |
| 3' end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16p; 3-alkyladenine DNA glycosylase; 3-methyladenine DNA glycosidase; 3-methyladenine DNA glycosylase; 9830006D05; AAG; ADPG; AI326268; alkylpurine-DNA-N-glycosylase; ANPG; APNG; CRA36.1; CRA36.1 (3-methyl-adenine DNA glycosylase); DNA-3-methyladenine glycosylase; MDG; Mid1; MPG; N-methylpurine DNA glycosylase; N-methylpurine-DNA glycocylase; N-methylpurine-DNA glycosylase; N-methylpurine-DNA glycosylase, MPG; PIG11; PIG16; proliferation-inducing protein 11; proliferation-inducing protein 16 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4350 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P29372 | |
| MPG | |
| Recombinant protein corresponding to Human MPG. Recombinant protein control fragment (Product #RP-89471). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction