missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPI Monoclonal Antibody (OTI4A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504771S
This item is not returnable.
View return policy
Description
MPI Monoclonal antibody specifically detects MPI in Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| MPI | |
| Monoclonal | |
| 0.9 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P35433, Q06203 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 117544, 231327, 5471, 611407 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI4A11 | |
| Unconjugated | |
| Ppat | |
| 1110002E17Rik; AI315153; CDG1B; mannose phosphate isomerase; mannose phosphate isomerase (mapped); mannose phosphate isomerase 1; mannose-6-phosphate isomerase; MPI; Mpi1; Mpi-1; Phosphohexomutase; phosphomannose isomerase; phosphomannose isomerase 1; PMI; PMI1 | |
| Human recombinant protein fragment corresponding to amino acids 42-278 of human PPAT produced in E.coli. | |
| 30 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction