missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mPlum Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-11833MFV500
This item is not returnable.
View return policy
Description
mPlum Polyclonal antibody specifically detects mPlum in Non-species specific samples. It is validated for Flow Cytometry
Specifications
| mPlum | |
| Polyclonal | |
| Flow Cytometry | |
| Fluorescent protein plum, mPlum Fluorescent Protein, Red Fluorescent Protein, RFP | |
| mPlum from Discosoma sp. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction