missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPO Mouse anti-Human, Clone: OTI6A9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813912S
This item is not returnable.
View return policy
Description
MPO Monoclonal antibody specifically detects MPO in Human samples. It is validated for ELISASpecifications
| MPO | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05164 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4353 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| OTI6A9 | |
| Unconjugated | |
| MPO | |
| 84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain | |
| Full length recombinant protein of human MPO (NP_000241) produced in Baculovirus-Insect cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction