missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPP10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536980
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 79 kDa and 120 kDa (phosphorylated). Purity is >95% by SDS-PAGE.
MPP10 (M-phase phosphoprotein 10), also known as MPHOSPH10, is a 681 amino acid protein that localizes to the fibrillar region of the nucleolus. Phosphorylated in the M (mitotic) phase of the cell cycle, MPP10 is a component of the 60-80S U3 small nucleolar ribonucleoprotein (U3 snoRNP) complex and is required for the early cleavages during pre-18S ribosomal RNA processing. MPP10 forms a heterotrimeric complex with IMP-3 and IMP-4, which may be required for its association with nucleolar components. The gene encoding MPP10 is localized to human chromosome 2, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, Neurofibromatosis type 2, autism and schizophrenia.
Specifications
| MPP10 | |
| Polyclonal | |
| Unconjugated | |
| MPHOSPH10 | |
| 2810453H10Rik; 5730405D16Rik; AI326008; CT90; M phase phosphoprotein 10; M-phase phosphoprotein 10; M-phase phosphoprotein 10 (U3 small nucleolar ribonucleoprotein); Mphosph10; MPP10; MPP10P; PPP1R106; protein phosphatase 1, regulatory subunit 106; U3 small nucleolar ribonucleoprotein; U3 small nucleolar ribonucleoprotein protein MPP10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10199, 293828, 67973 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00566, Q810V0 | |
| MPHOSPH10 | |
| Synthetic peptide corresponding to amino acids 126-171 of human MPP10. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction