missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPP3 Antibody (OTI8F1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72771F
This item is not returnable.
View return policy
Description
MPP3 Monoclonal specifically detects MPP3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| MPP3 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| MPP3 | |
| Full length recombinant protein of human MPP3 (NP_001923) produced in HEK293T cells. | |
| 0.1 mL | |
| 4356.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI8F1 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| Discs large homolog 3, discs, large homolog 3, DLG3MAGUK p55 subfamily member 3, membrane protein palmitoylated 3, membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3), Protein MPP3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction