missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPP3 Monoclonal Antibody (OTI8B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501306
This item is not returnable.
View return policy
Description
MPP3 is a member of a family of membrane-associated proteins termed MAGUKs (membrane-associated guanylate kinase homologs). MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. This protein contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction. Alternatively spliced transcript variants have been identified. One transcript variant is experimentally supported, but it doesn′t encode a protein.Specifications
| MPP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13368 | |
| MPP3 | |
| Full length human recombit protein of human MPP3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI8B11 | |
| Unconjugated | |
| MPP3 | |
| DLG3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4356 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction