missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPP3 Monoclonal Antibody (OTI8F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501303S
This item is not returnable.
View return policy
Description
MPP3 Monoclonal antibody specifically detects MPP3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| MPP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N4J0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 138199 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI8F1 | |
| Unconjugated | |
| CARNMT1 | |
| 6430514B01; CSG18; discs large homolog 3; discs, large homolog 3; DLG3; Dlgh3; Dusp3; MAGUK p55 subfamily member 3; MAGUK p55 subfamily member 3 {ECO:0000250; membrane palmitoylated protein 3; membrane protein palmitoylated 3; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3); Mpp3; mpp3 {ECO:0000312; Mpp3 membrane protein, palmitoylated 3; protein Dlgh3; protein MPP3; RGD:620015}; UniProtKB:O88910} | |
| Full length human recombinant protein of human C9orf41 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction