missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554519
This item is not returnable.
View return policy
Description
Immunogen sequence: VCLVDVEPEA LKQLRTSEFK PYIIFVKPAI QEKRKTPPMS PACEDTAAPF DEQQQEMAAS AAFIDRHYGH LVDAVLVKED LQGAYSQLKV VL Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.
This gene product is a member of a family of membrane-associated proteins termed MAGUKs (membrane-associated guanylate kinase homologs). MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. This protein contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction. Alternatively spliced transcript variants have been identified. One transcript variant is experimentally supported, but it doesn't encode a protein.
Specifications
| MPP3 | |
| Polyclonal | |
| Unconjugated | |
| Mpp3 | |
| 6430514B01; CSG18; discs large homolog 3; discs, large homolog 3; DLG3; Dlgh3; Dusp3; MAGUK p55 subfamily member 3; MAGUK p55 subfamily member 3 {ECO:0000250; membrane palmitoylated protein 3; membrane protein palmitoylated 3; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3); Mpp3; mpp3 {ECO:0000312; Mpp3 membrane protein, palmitoylated 3; protein Dlgh3; protein MPP3; RGD:620015}; UniProtKB:O88910} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4356 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13368 | |
| Mpp3 | |
| Recombinant protein corresponding to Human MPP3. Recombinant protein control fragment (Product #RP-93349). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction