missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599990
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MPP5.
MPP5 (Membrane Protein, Palmitoylated 5) plays a critical role in neurogenesis and hematopoiesis. This protein is crucial for establishing and maintaining cell polarity in epithelial cells and neurons. It is involved in the Par-3 complex, which contributes to maintaining tight junctions in epithelial cells. Mutations in MPP5 have been associated with severe neurologic impairments, including global developmental delay (GDD) and behavioral changes. In animal models, MPP5 is highly involved in the early differentiation of multipotent progenitors and supports long-term lineage-specific contributions, primarily in myeloid and lymphoid cells. MPP5 variants disrupt normal cellular function, leading to profound developmental and functional abnormalities.
Specifications
| MPP5 | |
| Polyclonal | |
| Unconjugated | |
| MPP5 | |
| 3830420B02Rik; AI255216; AI644496; FLJ12615; MAGUK p55 subfamily member 5; membrane palmitoylated protein 5; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 5); membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5); MPP5; palmitoylated 5; PALS1; protein associated with Lin Seven 1; protein associated with Lin-7 1; stardust | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 314259, 56217, 64398 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N3R9, Q9JLB2 | |
| MPP5 | |
| A synthesized peptide derived from human MPP5(Accession Q8N3R9), corresponding to amino acid residues S588-Y638. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction