missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558030
This item is not returnable.
View return policy
Description
Immunogen sequence: SSRDDQGAAK PFTEEDFQEM IKSAQIMESQ YGHLFDKIII NDDLTVAFNE LKTTFDKLET ETHWVPVSWL HS Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%.
Acts as an important adapter that promotes epithelial cell polarity and tight junction formation via its interaction with DLG1. Involved in the assembly of protein complexes at sites of cell-cell contact.
Specifications
| MPP7 | |
| Polyclonal | |
| Unconjugated | |
| MPP7 | |
| 1110068J02Rik; 2810038M04Rik; 5430426E14Rik; AI415104; Gm955; MAGUK p55 subfamily member 7; membrane palmitoylated protein 7; membrane protein, palmitoylated 7 (MAGUK p55 subfamily member 7); MPP7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 143098 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T2T1 | |
| MPP7 | |
| Recombinant protein corresponding to Human MPP7. Recombinant protein control fragment (Product #RP-97115). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction