missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mpp8 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71809V
Denna artikel kan inte returneras.
Se returpolicy
Description
mpp8 Polyclonal antibody specifically detects mpp8 in Human samples. It is validated for Western Blot
Specifications
| mpp8 | |
| Polyclonal | |
| Western Blot | |
| FLJ35237, HSMPP8, M-phase phosphoprotein 8, M-phase phosphoprotein, mpp, M-phase phosphoprotein, mpp8, MPP8, TWA3, Two hybrid-associated protein 3 with RanBPM | |
| The immunogen for this product maps to a region between residue 200 and 250 of human M-phase Phosphoprotein 8 using the numbering given in entry NP_059990.2 (GeneID 54737). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 54737 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction