missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPRIP Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 468.00
Specifications
| Antigen | MPRIP |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MPRIP Polyclonal antibody specifically detects MPRIP in Human samples. It is validated for Western BlotSpecifications
| MPRIP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 23164 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| KIAA0864Rho interacting protein 3, MRIPP116RIP, myosin phosphatase Rho interacting protein, p116Ripmyosin phosphatase-Rho interacting protein, RHOIP3myosin phosphatase Rho-interacting protein, RIP3M-RIPRho-interacting protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-310 of human MPRIP (NP_055949.2). MSAAKENPCRKFQANIFNKSKCQNCFKPRESHLLNDEDLTQAKPIYGGWLLLAPDGTDFDNPVHRSRKWQRRFFILYEHGLLRYALDEMPTTLPQGTINMNQCTDVVDGEGRTGQKFSLCILTPEKEHFIRAETKEIVSGWLEMLMVYPRTNKQNQKKKRKVEPPTPQEPGPAKVAVTSSSSSSSSSSSIPSAEKVPTTKSTLWQEEMRTKDQPDGSSLSPAQSPSQSQPPAASSLREPGLESKEEESAMSSDRMDCGRKVRVESGYFSLEKTKQDLKAEEQQLPPPLSPPSPSTPNHRRSQVIEKFEAL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title