missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Mpro 61

Description
Mpro 61 is a potent non-covalent inhibitor of SARS-CoV-2 main protease (IC50 = 44 nM, EC50 = 0.175 μM in the replicon assay and EC50 = 0.08 μM in the infectious SARS-CoV-2 assay); it is cell permable and exhibits no apparent cytotoxicity.
Specifications
Specifications
| Chemical Name or Material | 5-[5-[3-Chloro-5-[(2-chlorophenyl)methoxy]-4-fluorophenyl]-6-oxo[1(6 H),3 '-bipyridin]-3-yl]-1-methyl-2,4(1 H,3 H)-pyrimidinedione |
| CAS | 2679814-93-6 |
| Quantity | 5 mg |
| Target | 3 C and 3 CL Protease Inhibitors |
| Molecular Formula | C28H19Cl2FN4O4 |
| Purity | 0.95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?