missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPZ Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598291
This item is not returnable.
View return policy
Description
MPZ Polyclonal Antibody for Western Blot, ELISA
Zero, also known as myelin protein zero (MPZ) is a Type 1 integral membrane glycoprotein that mediates adhesion of spiraling wraps of the myelin sheath in order to ensure stable synaptic transmission. Zero protein encompasses approximately 50% of total protein in the sheath scaffolding in contribution to structural integrity of peripheral myelin. Zero guides the compact myelin wrapping process through glycine zipper packing interface-dependent dimer and tetramer formation. Mutations (e.g. G134R) can abrogate multimer formation, cause demyelinating neuropathies, and are known to contribute to conditions that include Charcot-Marie-Tooth disease. Zero cytoplasmic domain undergoes serine and tyrosine phosphorylation, which appears to be prevalent during peak nerve myelination. Zero transcript is moderate in brain, abundant in thymus and most abundant in white matter of the CNS.
Specifications
| MPZ | |
| Polyclonal | |
| Unconjugated | |
| MPZ | |
| Charcot-Marie-Tooth neuropathy 1B; CHM; CMT1; CMT1B; CMT2I; CMT2J; CMT4E; CMTDI3; CMTDID; DSS; HMSNIB; MPP; MPZ; Myelin peripheral protein; myelin protein P0; Myelin protein zero; myelin protein zero (Charcot-Marie-Tooth neuropathy 1B); P0; P-zero | |
| Rabbit | |
| Protein G | |
| RUO | |
| 4359 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P25189 | |
| MPZ | |
| Recombinant Human Myelin protein P0 protein (30-156AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction