missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPZL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 292.00 - € 572.00
Specifications
| Antigen | MPZL1 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:10-1:20 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18439500
|
Novus Biologicals
NBP1-83625-25ul |
25 μL |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18260207
|
Novus Biologicals
NBP1-83625 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MPZL1 Polyclonal specifically detects MPZL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MPZL1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| FLJ21047, immunoglobulin family transmembrane protein, MPZL1b, myelin protein zero-like 1, myelin protein zero-like protein 1, protein zero related, Protein zero-related, PZR1b, PZRb, PZRPZRa | |
| MPZL1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:10-1:20 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9019 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KEIFVANGTQGKLTCKFKSTSTTGGLTSVSWSFQPEGADTTVSFFHYSQGQVYLGNYPPFKDRISWAGDLDKKDASINIENMQFIHNGTYICDVKNPPDIVVQPGHIRLYVVEKENLPVFPVWVV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title