missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPZL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11787-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can be used to detect endogenous MPZL2, however, the reason for the additional band of 32 kDa is still unknown.
Epithelial V-like antigen (EVA) is expressed in thymus epithelium and strongly downregulated by thymocyte developmental progression. This gene is expressed in the thymus and in several epithelial structures early in embryogenesis.Specifications
| MPZL2 | |
| Polyclonal | |
| Unconjugated | |
| MPZL2 | |
| Epithelial V like antigen 1, EVA, EVA1, MPZL2, myelin protein zero like 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10205, 14012, 300679 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60487, O70255 | |
| MPZL2 | |
| MPZL2 Fusion Protein Ag2419 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction