missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MR1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 201.00 - € 482.00
Specifications
| Antigen | MR1 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682711
|
Novus Biologicals
NBP2-93753-0.02ml |
0.02 mL |
€ 201.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616671
|
Novus Biologicals
NBP2-93753-0.1ml |
0.1 mL |
€ 482.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MR1 Polyclonal antibody specifically detects MR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| MR1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS (pH 7.3), 50% glycerol | |
| 3140 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Class I histocompatibility antigen-like protein, FLJ31593, HLALS, major histocompatibility complex class I-related gene protein, major histocompatibility complex, class I-like sequence, major histocompatibility complex, class I-related, MHC class I-like antigen MR-1, MHC class I-related gene protein, MHC class-I related-gene protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 23-291 of human MR1 (NP_001522.1). RTHSLRYFRLGVSDPIHGVPEFISVGYVDSHPITTYDSVTRQKEPRAPWMAENLAPDHWERYTQLLRGWQQMFKVELKRLQRHYNHSGSHTYQRMIGCELLEDGSTTGFLQYAYDGQDFLIFNKDTLSWLAVDNVAHTIKQAWEANQHELLYQKNWLEEECIAWLKRFLEYGKDTLQRTEPPLVRVNRKETFPGVTALFCKAHGFYPPEIYMTWMKNGEEIVQEIDYGDILPSGDGTYQAWASIELDPQSSNLYSCHVEHCGVHMVLQV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title