missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MR1 Antibody (MR1/7579) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26945V
This item is not returnable.
View return policy
Description
MR1 Monoclonal antibody specifically detects MR1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MR1 | |
| Monoclonal | |
| DyLight 405 | |
| Class I histocompatibility antigen-like protein, FLJ31593, HLALS, major histocompatibility complex class I-related gene protein, major histocompatibility complex, class I-like sequence, major histocompatibility complex, class I-related, MHC class I-like antigen MR-1, MHC class I-related gene protein, MHC class-I related-gene protein | |
| Recombinant fragment (around aa50-250) of human MR1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Immunology | |
| 3140 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| MR1/7579 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction