missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRAS Mouse anti-Human, Clone: OTI3C4, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502864
This item is not returnable.
View return policy
Description
MRAS is a member of the RAS superfamily of GTP-binding proteins which are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. MRAS is oncogenically activated in a significant fraction of tumors.Specifications
| MRAS | |
| Monoclonal | |
| 0.69 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14807 | |
| MRAS | |
| Full length human recombit protein of human MRAS produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3C4 | |
| Unconjugated | |
| MRAS | |
| M-RAs, R-RAS3, RRAS3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 22808 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction