missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRAS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571377
This item is not returnable.
View return policy
Description
MRAS Polyclonal Antibody for Western Blot
MRAS is a member of the RAS superfamily of GTP-binding proteins which are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. MRAS is oncogenically activated in a significant fraction of tumors.
Specifications
| MRAS | |
| Polyclonal | |
| Unconjugated | |
| MRAS | |
| 2900078C09Rik; AI326250; MRAS; M-RAs; Muscle and microspikes Ras; muscle RAS oncogene homolog; ras-related protein M-Ras; Ras-related protein R-Ras3; RRAS3; R-RAS3; Xras; X-Ras | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17532 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O08989 | |
| MRAS | |
| synthetic peptide directed towards the C terminal region of human MRAS. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction