missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRAS Recombinant Rabbit Monoclonal Antibody (3F6Q10)

Product Code. 30132042
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132042 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30132042 Supplier Invitrogen™ Supplier No. MA555490

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen Sequence: LILRVKDRES FPMILVANKV DLMHLRKITR EQGKEMATKH NIPYIETSAK DPPLNVDKAF HDLVRVIRQQ IPEKSQKKKK KTKWRGDRAT GTHKLQCVIL.

MRAS is a member of the RAS superfamily of GTP-binding proteins which are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. MRAS is oncogenically activated in a significant fraction of tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRAS
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3F6Q10
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MRAS
Gene Accession No. O08989, O14807, P97538
Gene Alias 2900078C09Rik; AI326250; MRAS; M-RAs; Muscle and microspikes Ras; muscle RAS oncogene homolog; ras-related protein M-Ras; Ras-related protein R-Ras3; RRAS3; R-RAS3; Xras; X-Ras
Gene Symbols MRAS
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 109-208 of human Mras.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17532, 22808, 25482
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.