missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Mrc2 Antibody (Quark Patent Anti-Endo180), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28131-50ug
This item is not returnable.
View return policy
Description
Mrc2 Monoclonal antibody specifically detects Mrc2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Mrc2 | |
| Monoclonal | |
| Unconjugated | |
| Q9UBG0 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Quark Patent Anti-Endo180 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD280 antigen, CD280C-type mannose receptor 2, CLEC13EUPAR-associated protein, ENDO180C-type lectin domain family 13 member E, Endocytic receptor 180, KIAA0709FLJ35911, Macrophage mannose receptor 2, mannose receptor, C type 2, UPARAPendocytic receptor (macrophage mannose receptor family), urokinase plasminogen activator receptor-associated protein, Urokinase receptor-associated protein, Urokinase-type plasminogen activator receptor-associated protein | |
| MRC2/ CD280 | |
| 50 μg | |
| CD Markers, Cell Biology, Immunology | |
| 9902 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction