missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRCK Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-46822F
This item is not returnable.
View return policy
Description
MRCK Polyclonal antibody specifically detects MRCK in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| MRCK | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| CDC42 binding protein kinase alpha (DMPK-like), CDC42 binidng protein kinase beta, CDC42-binding protein kinase alpha, CDC42-binding protein kinase alpha (DMPK-like), DMPK-like alpha, EC 2.7.11, EC 2.7.11.1, KIAA0451DKFZp686L1738, MRCK, MRCK alpha, MRCKADKFZp686P1738, Myotonic dystrophy kinase-related CDC42-binding kinase alpha, myotonic dystrophy kinase-related CDC42-binding protein kinase alpha, Myotonic dystrophy protein kinase-like alpha, PK428FLJ23347, serine/threonine-protein kinase MRCK alpha, ser-thr protein kinase PK428, ser-thr protein kinase related to the myotonic dystrophy protein kinase | |
| The immunogen for this product maps to a region between residue 1560 and 1610 of human myotonic dystrophy protein kinase-like alpha using the numbering given in entry CAH71185.1 (GeneID 8476). | |
| 0.1 mL | |
| Protein Kinase | |
| 8476 | |
| Store at 4C in the dark. | |
| IgG |
| Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction