missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mre11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 199.00 - € 351.00
Specifications
| Antigen | Mre11 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 20802516), Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18086464
|
Novus Biologicals
NB100-56339SS |
0.025 mg |
€ 199.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18096825
|
Novus Biologicals
NB100-56339 |
0.1 mg |
€ 351.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mre11 Polyclonal specifically detects Mre11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Mre11 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 20802516), Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| NP_005581 | |
| 4361 | |
| Mre11 Antibody was made to a synthetic peptide of human MRE11 used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase | |
| PBS with 0.02% Sodium Azide | |
| ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B | |
| MRE11 | |
| IgG | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title