missing translation for 'onlineSavingsMsg'
Learn More

MRFAP1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 190.00 - € 470.00

Specifications

Antigen MRFAP1
Dilution Western Blot 1:200-1:1000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18670902
View Documents
Novus Biologicals
NBP2-94813-0.02ml
0.02 mL
€ 190.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18623392
View Documents
Novus Biologicals
NBP2-94813-0.1ml
0.1 mL
€ 470.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MRFAP1 Polyclonal antibody specifically detects MRFAP1 in Human samples. It is validated for Western Blot
Specifications

Specifications

MRFAP1
Western Blot
Unconjugated
Rabbit
Cell Biology
PBS (pH 7.3), 50% glycerol
93621
IgG
Affinity purified
Western Blot 1:200-1:1000
Polyclonal
Purified
RUO
Human
Mof4 family associated protein 1, Morf4 family associated protein 1, PAM14MORF4 family-associated protein 1, PGR1protein associated with MRG, 14 kDa, Protein associated with MRG of 14 kDa, Protein PGR1, T-cell activation protein
Recombinant fusion protein containing a sequence corresponding to amino acids 1-127 of human MRFAP1 (NP_150638.1). MRPLDIVELAEPEEVEVLEPEEDFEQFLLPVINEMREDIASLTREHGRAYLRNRSKLWEMDNMLIQIKTQVEASEESALNHLQNPGDAAEGRAAKRCEKAEEKAKEIAKMAEMLVELVRRIEKSESS
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.