missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRG15 Antibody (OTI5D2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72778DL594
This item is not returnable.
View return policy
Description
MRG15 Monoclonal antibody specifically detects MRG15 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MRG15 | |
| Monoclonal | |
| DyLight 594 | |
| Eaf3, Esa1p-associated factor 3 homolog, HsT17725, MEAF3, MGC10631, MORF-related gene on chromosome 15, MORFRG15, mortality factor 4 like 1, mortality factor 4-like protein 1, MRG15MORF-related gene 15 protein, Protein MSL3-1, S863-6, Transcription factor-like protein MRG15 | |
| Recombinant protein fragment corresponding to amino acids 6-77 of human MRG15 (NP_006782) produced in E.coli. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 10933 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5D2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction