missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRGPRF Antibody (916219) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8396J
This item is not returnable.
View return policy
Description
MRGPRF Monoclonal antibody specifically detects MRGPRF in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| MRGPRF | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 916219 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| FLJ16111, FLJ29034, FLJ40998, FLJ53714, G protein-coupled receptor 140, G protein-coupled receptor 168, G protein-coupled receptor MrgF, GPR140, GPR168, G-protein coupled receptor 140, G-protein coupled receptor 168, mas-related G protein-coupled MRGF, Mas-related gene F protein, MAS-related GPR, member F, mas-related G-protein coupled receptor member F, MGC21621, mrgF, MRGFDKFZp586B2122, RTA, seven transmembrane helix receptor | |
| NS0 mouse myeloma cell line transfected with human MRGPRF, Met1-Ser343, Accession # NP_659452 | |
| 0.1 mL | |
| GPCR | |
| 116535 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction