missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRGPRF Polyclonal Antibody
GREENER_CHOICE

Product Code. 13444599
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13444599 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13444599 Supplier Invitrogen™ Supplier No. PA532929

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%); Marmoset, Dog, Panda, Pig (94%); Bovine, Horse (89%); Elephant (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

May bind to a neuropeptide and may regulate nociceptor function and/or development, including the sensation or modulation of pain. Expressed in the Gut, vas deferens, uterus and aorta; barely detectable in liver, kidney, lung, and salivary gland. In the brain, markedly abundant in the cerebellum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRGPRF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MRGPRF
Gene Accession No. Q96AM1
Gene Alias BC019711; G protein coupled receptor (RTA); G protein coupled receptor Rca; G protein-coupled receptor 140; G protein-coupled receptor 168; G protein-coupled receptor MrgF; G protein-coupled receptor RTA; G-p; GPR140; GPR168; G-protein coupled receptor 140; G-protein coupled receptor 168; G-protein coupled receptor RTA; MAS related GPR family member F; mas-related G protein-coupled MRGF; Mas-related G protein-coupled receptor c; mas-related gene; mas-related gene F protein; MAS-related GPR, member F; mas-related G-protein coupled receptor member F; MAS-related G-protein coupled receptor, member F; MGC21621; Mgrc; MRGF; MRGPRF; PSEC0142; Rta; seven transmembrane helix receptor
Gene Symbols MRGPRF
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from C-terminal cytoplasmic domain of human MRGPRF.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116535
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.