missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRGX3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS5120
This item is not returnable.
View return policy
Description
MRGX3 Polyclonal antibody specifically detects MRGX3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MRGX3 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 21 μg/mL | |
| G protein-coupled receptor MRGX3, G protein-coupled receptor SNSR2, GPCR, mas-related GPCR member X3, MAS-related GPR, member X3, mas-related G-protein coupled receptor member X3, MRGX3G protein-coupled receptor SNSR1, Sensory neuron-specific G-protein coupled receptor 1/2, SNSR1, SNSR2 | |
| Synthetic 17 amino acid peptide from 1st extracellular domain of human MRGX3. | |
| 0.05 mL | |
| GPCR | |
| 117195 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction