missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRLC2 Antibody (SR1293), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22501-50ul
This item is not returnable.
View return policy
Description
MRLC2 Monoclonal antibody specifically detects MRLC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| MRLC2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR1293 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| MLC-2, MLC20, MLC-2A, MLC-B, MRLC2SHUJUN-1, MYLC2B, myosin regulatory light chain 12B, myosin regulatory light chain 2, Myosin regulatory light chain 20 kDa, Myosin regulatory light chain 2-B, smooth muscle isoform, Myosin regulatory light chain MRLC2, myosin, light chain 12B, regulatory | |
| A synthesized peptide derived from human MRLC2 (Uniprot #: O14950) | |
| 50 μg | |
| Cell Biology, Cytoskeleton Markers | |
| 103910 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto