missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP1 Antibody (MRP1/1344), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08228JF549
This item is not returnable.
View return policy
Description
MRP1 Monoclonal antibody specifically detects MRP1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| MRP1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein | |
| Recombinant full-length human MRP1 protein (Uniprot: P33527) | |
| 100 μg | |
| ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
| 4363 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, ELISA, Immunofluorescence | |
| MRP1/1344 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction