missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP4/ABCC4 Antibody (ABCC4/9019), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23893-20ug
This item is not returnable.
View return policy
Description
MRP4/ABCC4 Monoclonal antibody specifically detects MRP4/ABCC4 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MRP4/ABCC4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| ATP-binding cassette sub-family C member 4, ATP-binding cassette, sub-family C (CFTR/MRP), member 4, EST170205, MOAT-BbA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4), MOATBcanalicular multispecific organic anion transporter (ABC superfamily), MRP4MRP/cMOAT-related ABC transporter, multidrug resistance-associated protein 4, multispecific organic anion transporter B, Multi-specific organic anion transporter B | |
| Recombinant fragment (around aa1-200) of human MRP4/ABCC4 protein (exact sequence is proprietary) | |
| 20 μg | |
| Lipid and Metabolism | |
| 10257 | |
| Store at 4°C. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| ABCC4/9019 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction