missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRP6 Monoclonal Antibody (M6II-31)
GREENER_CHOICE

Product Code. 11579522
Change view
Click to view available options
Quantity:
250 μL
Unit Size:
250µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11579522 250 μL 250µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11579522 Supplier Invitrogen™ Supplier No. MA126542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Recommended positive controls: Liver, kidney and cell Lines (drug-sensitive parental cell lines and their multidrug-resistant derivatives)..

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). The encoded protein, a member of the MRP subfamily, is involved in multi-drug resistance. Mutations in this gene cause pseudoxanthoma elasticum. Alternatively spliced transcript variants that encode different proteins have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRP6
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone M6II-31
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 1% BSA and 0.1% sodium azide
Gene ABCC6
Gene Accession No. O95255
Gene Alias ABC34; Abcc1b; ABCC6; Anthracycline resistance-associated protein; ARA; ATP binding cassette subfamily C member 6; ATP-binding cassette sub-family C member 6; ATP-binding cassette, subfamily C (CFTR/MRP), member 6; ATP-binding cassette, sub-family C (CFTR/MRP), member 6; ATP-binding cassette, sub-family C, member 6; DCC; dyscalc; Dyscalc1; EST349056; GACI2; liver multidrug resistance-associated protein 6; Mlp1; MLP-1; MOATE; MOAT-E; Mrp6; MRP-like protein 1; Multidrug resistance-associated protein 6; multidrug resistance-associated protein-6; multi-specific organic anion transporter E; PXE; PXE1; URG7; URG7 protein; multidrug resistance-associated protein 6
Gene Symbols ABCC6
Host Species Rat
Immunogen Bacterial fusion protein of human MRP 6, containing amino acids 764-964, spanning the putative 12th transmembrane region as well as predicted internal and external regions of the protein.
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 368
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.