missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. p-200128834
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17730694 200 μL 200µL
17733164 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17730694 Supplier Invitrogen™ Supplier No. PA5119287

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MRPL1 Polyclonal Antibody for IHC (P), Western Blot

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein that belongs to the L1 ribosomal protein family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MRPL1
Gene Accession No. Q9BYD6
Gene Alias 2410002L03Rik; 39S ribosomal protein L1, mitochondrial; 5830418D04Rik; AI462572; BM022; BM-022; L1mt; Mitochondrial large ribosomal subunit protein uL1m; mitochondrial ribosomal protein L1; Mrpl1; MRP-L1
Gene Symbols MRPL1
Host Species Rabbit
Immunogen Recombinant protein Mitochondrial Ribosomal Protein L1. The antigen corresponds to amino acid range 147-285 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65008
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.