missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563907
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Immunogen sequence: HLRDPVAIVK LAIYGMLPKN LHRRTMMERL HLFPDEYIPE DILKNLVEEL PQPRKIPKRL DEYTQEEIDA FPRLWTPPE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Tekniske data
| MRPL13 | |
| Polyclonal | |
| Unconjugated | |
| MRPL13 | |
| 1110002D09Rik; 39S ribosomal protein L13, mitochondrial; L13; L13A; L13mt; Mitochondrial large ribosomal subunit protein uL13m; mitochondrial ribosomal protein L13; Mrpl13; MRP-L13; RPL13; RPML13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28998 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.7 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BYD1 | |
| MRPL13 | |
| Recombinant protein corresponding to Human MRPL13. Recombinant protein control fragment (Product #RP-101966). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion