missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551007
This item is not returnable.
View return policy
Description
MRPL13 Polyclonal Antibody detects endogenous levels of MRPL13 protein.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Specifications
| MRPL13 | |
| Polyclonal | |
| Unconjugated | |
| MRPL13 | |
| 1110002D09Rik; 39S ribosomal protein L13, mitochondrial; L13; L13A; L13mt; Mitochondrial large ribosomal subunit protein uL13m; mitochondrial ribosomal protein L13; Mrpl13; MRP-L13; RPL13; RPML13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28998, 299938, 68537 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BYD1, Q9D1P0 | |
| MRPL13 | |
| Synthesized peptide derived from the C-terminal region of human MRP-L13. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction