missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL15 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 468.00
Specifications
| Antigen | MRPL15 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MRPL15 Polyclonal antibody specifically detects MRPL15 in Human, Mouse samples. It is validated for Western BlotSpecifications
| MRPL15 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 29088 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| L15mtHSPC145, mitochondrial ribosomal protein L15, MRP-L1539S ribosomal protein L15, mitochondrial, MRP-L7, RPML7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-296 of human MRPL15 (NP_054894.1). MAGPLQGGGARALDLLRGLPRVSLANLKPNPGSKKPERRPRGRRRGRKCGRGHKGERQRGTRPRLGFEGGQTPFYIRIPKYGFNEGHSFRRQYKPLSLNRLQYLIDLGRVDPSQPIDLTQLVNGRGVTIQPLKRDYGVQLVEEGADTFTAKVNIEVQLASELAIAAIEKNGGVVTTAFYDPRSLDIVCKPVPFFLRGQPIPKRMLPPEELVPYYTDAKNRGYLADPAKFPEARLELARKYGYILPDITKDELFKMLCTRKDPRQIFFGLAPGWVVNMADKKILKPTDENLLKYYTS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title