missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL21 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562451
This item is not returnable.
View return policy
Description
Immunogen sequence: ACSHSILRPS GPGAASLWSA SRRFNSQSTS YLPGYVPKTS LSSPPWPEVV LPDPVEETRH HAEVV Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 82%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Multiple transcript variants encoding different isoforms were identified through sequence analysis although some may be subject to nonsense-mediated decay (NMD).
Specifications
| MRPL21 | |
| Polyclonal | |
| Unconjugated | |
| MRPL21 | |
| 39S ribosomal protein L21, mitochondrial; BC028768; D9Wsu149; L21mt; Mitochondrial large ribosomal subunit protein bL21m; mitochondrial ribosomal protein L21; Mrpl21; MRP-L21; Unknown (protein for MGC:133585) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 219927 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z2W9 | |
| MRPL21 | |
| Recombinant protein corresponding to Human MRPL21. Recombinant protein control fragment (Product #RP-100632). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction