missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL23 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06349-50ul
This item is not returnable.
View return policy
Description
MRPL23 Polyclonal antibody specifically detects MRPL23 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MRPL23 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| FLJ45387, L23MRPmitochondrial, mitochondrial ribosomal protein L23 | |
| Produced in rabbits immunized with E. coli-derived Human MRPL23 fragment. | |
| 50 μg | |
| Endocrinology | |
| 6150 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction